missing translation for 'onlineSavingsMsg'
Learn More

TRA-1-81 Antigen Mouse anti-Human, PE, Clone: TRA-1-81, BD

Mouse Monoclonal Antibody

67.25€ - 295.87€

Specifications

Antigen TRA-1-81 Antigen
Clone TRA-1-81
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15820569
View Documents
BD Biosciences
560885
25 Tests
ONLINE EXCLUSIVE PRICE
67.45€
67.25€ / Each
Save 0.20€
0.30% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15839978
View Documents
BD Biosciences
560161
100 Tests
ONLINE EXCLUSIVE PRICE
296.00€
295.87€ / Each
Save 0.13€
0.04% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The TRA-1-81 monoclonal antibody reacts with a pluripotent-stem-cell-specific epitope on a high-molecular-weight transmembrane glycoprotein. The TRA-1-81 antigen is an epitope on the same keratan sulfate core molecule, podocalyxin, as 4 other distinct antigens on tumor-derived cell lines, TRA-1-60, GCTM2, K4, and K21. The expression of TRA-1-81 antigen is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. The antigen is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (but not morula or trophoblast), and embryonic stem (ES) cells. As human EC and ES cells undergo differentiation, expression of TRA-1-81 antigen is lost.

Specifications

Specifications

TRA-1-81 Antigen
Flow Cytometry
PE
RUO
Human
Human Embryonal Carcinoma Cell Line
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
TRA-1-81
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgM κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.