Learn More
Invitrogen™ TRA-1-81 (Podocalyxin) Monoclonal Antibody (TRA-1-81), PE, eBioscience™

Mouse Monoclonal Antibody
294.00€
Specifications
| Antigen | TRA-1-81 (Podocalyxin) |
|---|---|
| Clone | TRA-1-81 |
| Concentration | 0.2 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: The TRA-1-81 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-81 antibody is lost upon cell differentiation. The TRA-1-81 epitope is resistant to neuraminidase digestion, unlike the epitope recognized by the related TRA-1-60 antibody. The TRA-1-81 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer. Applications Reported: This TRA-1-81 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TRA-1-81 antibody has been tested by flow cytometric analysis of the human embryonal carcinoma (EC) line 2102Ep. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of inter...
TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.Specifications
| TRA-1-81 (Podocalyxin) | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Primary | |
| Affinity chromatography |
| TRA-1-81 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgM | |
| Human | |
| GCTM-2 antigen; Gp200; PC; PCLP; PCLP1; PCLP-1; Podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; PODXL; TRA180; tra1-81 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.