missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAF3 Isoform 2 Monoclonal Antibody (433113)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561287
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
TRAF3 Isoform 2 is a variant of the TNF receptor-associated factor 3 (TRAF3) gene, which plays a crucial role in the regulation of immune responses, inflammation, and cell survival. TRAF3 is a part of the TRAF protein family, which mediates signal transduction from various receptors of the TNF receptor superfamily, Toll-like receptors (TLRs), and other immune receptors. Isoform 2 of TRAF3 may have distinct regulatory functions compared to the canonical form, potentially influencing specific signaling pathways. TRAF3 is known to act as a negative regulator of the non-canonical NF-kappaB signaling pathway, which is important for immune homeostasis and prevention of chronic inflammation. Alterations or mutations in TRAF3, including its isoforms, have been associated with various diseases, including autoimmune disorders, lymphomas, and other cancers. Research into TRAF3 Isoform 2 could provide insights into its unique roles and therapeutic potential in modulating immune responses and treating related diseases.
Specifications
| TRAF3 Isoform 2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| E. coli-derived recombinant human TRAF-3, Met1-Pro543. | |
| 250 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 433113 | |
| Unconjugated | |
| Rat | |
| Protein A/G | |
| RUO | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction