missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11780-1-AP-20UL
This item is not returnable.
View return policy
Description
TRAFs are signal transducers for members of TNFR superfamily. These are critically involved in activation of MAP3K family members involved in NF-kappaB and JNK activation and apoptosis. TRAF7, a novel 670 A. A protein, is a human homolog of mouse Traf7 andconsists of an N-terminal RING finger domain, central TRAF domain, a coiled-coil motif and 7 WD40 repeats at the C-terminus. It has E3-ubiquitin ligase activity for self-ubiquitination. TRAF7 specifically interacts with MEKK3 and mediates MEKK3-induced AP1 and CHOP activation and induces apoptosis through Caspase-independent pathways. It is a multifunctional protein involved in sumoylation and negative regulation of c-Myb, thereby regulating proliferation and differentiation of hematopoietic cells.Specifications
| TRAF7 | |
| Polyclonal | |
| Unconjugated | |
| TRAF7 | |
| RFWD1, RING finger protein 119, RNF119, TRAF7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 224619, 360491, 84231 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6Q0C0, Q922B6 | |
| Traf7 | |
| TRAF7 Fusion Protein Ag2414 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction