missing translation for 'onlineSavingsMsg'
Learn More

Tragacanth Gum MP Biomedicals

Product Code. 11457621 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
11457621 1 kg 1kg
11437621 100 g 100g
11447621 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11457621

Brand: MP Biomedicals 0210479291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9000-65-1
MDL Number MFCD00131255
Synonym Gum tragacanth

Specifications

Color Cream
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Tragacanth Gum, Molecular biology and Cell Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.