missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRANCE/TNFSF11/RANK L Antibody Pair [HRP]

Product Code. 18623640 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18623640 5 Plates Pack of 5
18263648 15 Plates Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18623640 Supplier Novus Biologicals™ Supplier No. NBP2793575Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRANCE/TNFSF11/RANK L.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 234.38 pg/mL
Assay Sensitivity 234.38-15000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRANCE/TNFSF11/RANK L.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Gene ID (Entrez) 8600
Gene Symbol TNFSF11
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human TRANCE/TNFSF11/RANK L monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Human TRANCE/TNFSF11/RANK L monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 570 ng of recombinant Human TRANCE/TNFSF11/RANK L.
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target TRANCE/TNFSF11/RANK L
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 15000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.