missing translation for 'onlineSavingsMsg'
Learn More

Tranilast, TRC

Product Code. 30455758
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455758 50 mg 50mg
30452290 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455758 Supplier Toronto Research Chemicals Supplier No. TRCT71457050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53902-12-8
Molecular Formula C18 H17 N O5
Molecular Weight (g/mol) 327.33
Synonym Tranilast - Bio-X TM
IUPAC Name 2-[[(E)-3-(3,4-dimethoxyphenyl)prop-2-enoyl]amino]benzoic acid
SMILES COc1ccc(\C=C\C(=O)Nc2ccccc2C(=O)O)cc1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H17NO5/c1-23-15-9-7-12(11-16(15)24-2)8-10-17(20)19-14-6-4-3-5-13(14)18(21)22/h3-11H,1-2H3,(H,19,20)(H,21,22)/b10-8+
Formula Weight 327.1107
Chemical Name or Material Tranilast
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.