missing translation for 'onlineSavingsMsg'
Learn More

trans-(15S)-Latanoprost, TRC

Product Code. 30467330
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467330 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467330 Supplier Toronto Research Chemicals Supplier No. TRCL1773050.25MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans (15S)-Latanoprost
CAS 1235141-39-5
Molecular Formula C26 H40 O5
Quantity 0.25 mg
Synonym (5E)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]-5-heptenoic Acid 1-Methylethyl Ester, Isoproyl (E)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]-5-heptenoate, (5E,15S)-Latanoprost, 5,6-trans-(15S)-Latanoprost
InChI Formula InChI=1S/C26H40O5/c1-19(2)31-26(30)13-9-4-3-8-12-22-23(25(29)18-24(22)28)17-16-21(27)15-14-20-10-6-5-7-11-20/h3,5-8,10-11,19,21-25,27-29H,4,9,12-18H2,1-2H3/b8-3+/t21-,22-,23-,24+,25-/m1/s1
SMILES CC(C)OC(=O)CCC\C=C\C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1CC[C@H](O)CCc2ccccc2
IUPAC Name propan-2-yl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate
Molecular Weight (g/mol) 432.59
Formula Weight 432.2876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.