missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Butenyl acetate, 95%

Product Code. 11382237
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382237 5 g 5g
11392237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382237 Supplier Thermo Scientific Alfa Aesar Supplier No. L09734.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 628-08-0 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-08-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00039912
InChI Key WNHXJHGRIHUOTG-ONEGZZNKSA-N
Synonym trans-2-butenyl acetate, crotyl acetate, e-2-butenyl acetate, 2-buten-1-ol, acetate, e-crotyl acetate, e-but-2-enyl acetate, 2-buten-1-ol, acetate, 2e, 2-buten-1-ol, 1-acetate, 2e, 2e-but-2-en-1-yl acetate, e-2-buten-1-yl acetate
PubChem CID 5363082
IUPAC Name [(E)-but-2-enyl] acetate
SMILES CC=CCOC(=O)C

Specifications

Density 0.919
Boiling Point 128°C to 130°C
Flash Point 31°C (87°F)
Refractive Index 1.42
Quantity 5 g
UN Number UN3272
Beilstein 1720994
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material trans-2-Butenyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.