missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexaneacetic acid, 97%

Product Code. 15408648
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408648 5 g 5g
15456976 250 mg 250mg
15498638 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408648 Supplier Thermo Scientific Alfa Aesar Supplier No. H62058.06
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 189153-10-4 | C13H23NO4 | 257.33 g/mol

trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water (1.1 g/L) (25°C)

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189153-10-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD03844588
InChI Key IHXBNSUFUFFBRL-UHFFFAOYSA-N
Synonym 2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
PubChem CID 11253897
IUPAC Name 2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC(=O)O

Specifications

Solubility Information Slightly soluble in water (1.1g/L) (25°C)
Quantity 5 g
Formula Weight 257.32
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexaneacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.