missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline, TRC

Product Code. 30135610
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135610 25 g 25g
30137785 100 g 100g
30003424 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135610 Supplier Toronto Research Chemicals Supplier No. TRCH95237625G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5 H9 N O3
Molecular Weight (g/mol) 131.13
Synonym (4R)-4-Hydroxy-L-proline, L-Proline, 4-hydroxy-, trans-, Proline, 4-hydroxy-, L- (8CI), (4R)-4-Hydroxy-L-proline, (-)-4-Hydroxy-2-pyrrolidinecarboxylic acid, (2S,4R)-(-)-4-Hydroxyproline, (2S,4R)-4-Hydroxyproline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, (R)-4-Hydroxy-(S)-proline, (R)-4-Hydroxy-L-proline, (S)-Hydroxyproline, 4(R)-Hydroxy-2(S)-pyrrolidinecarboxylic acid, 4(R)-Hydroxyproline, 4-Hydroxy-(S)-proline, 4-Hydroxy-L-proline, 4-Hydroxyproline, 4-trans-Hydroxy-L-proline, Hydroxy-L-proline, Hydroxyproline, Hypro, L-4-Hydroxyproline, L-Hydroxyproline, L-Hypro, L-trans-4-Hydroxyproline, Ls-Hydroxyproline, NSC 46704, Proline, 4-hydroxy-, trans-4-Hydroxy-L-proline, trans-4-Hydroxyproline, trans-L-4-Hydroxyproline, trans-L-Hydroxyproline, δ-Hydroxyproline
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CN[C@@H](C1)C(=O)O

Specifications

InChI Formula InChI=1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1
Quantity 25 g
Formula Weight 131.0582
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Hydroxyproline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.