missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Phenyl-3-buten-2-one, TRC

Product Code. 30420375
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420375 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420375 Supplier Toronto Research Chemicals Supplier No. TRCP319580250MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1896-62-4
Molecular Formula C10H10O
Quantity 250 mg
Synonym (3E)-4-Phenyl-3-buten-2-one, (E)-4-Phenyl-3-buten-2-one, (E)-Benzalacetone, (E)-Benzylideneacetone, (E)-Methyl Styryl Ketone, Methyl (E)-2-Phenylethenyl Ketone, Methyl trans-Styryl Ketone, trans-1-Phenylbut-1-en-3-one, trans-4-Phenyl-3-buten-2-one, trans-4-Phenyl-3-butene-2-one, trans-Benzalacetone, trans-Benzylideneacetone, trans-Phenylvinyl Methyl Ketone
InChI Formula InChI=1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+
SMILES CC(=O)\C=C\c1ccccc1
IUPAC Name (E)-4-phenylbut-3-en-2-one
Molecular Weight (g/mol) 146.19
Formula Weight 146.0732
Chemical Name or Material trans-4-Phenyl-3-buten-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.