Learn More
trans-Anethole, 99%
Product Code. p-2692361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10149931 5 mL Glass bottle 5mL
10081850 100 mL Glass bottle 100mL
10222942 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10149931 Supplier Thermo Scientific Acros Supplier No. 152530050
Add to basket
CAS: 4180-23-8 | C10H12O | 148.2 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC
Specifications
Melting Point 23.0°C
Color Colorless to Yellow
Density 0.9875g/mL
Boiling Point 234.0°C to 237.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHC6H4OCH3
Refractive Index 1.5590 to 1.5620
Quantity 5 mL
Beilstein 06, 566
Merck Index 15, 636
Specific Gravity 0.9875
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with ether, chloroform, soluble in benzene, ethyl acetate, acetone,, alcohol and carbon disulfide
Formula Weight 148.2
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material trans-Anethole, 99%