missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamamide, TRC

Product Code. 30367899
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367899 10 g 10g
30367122 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367899 Supplier Toronto Research Chemicals Supplier No. TRCC44202510G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22031-64-7
Molecular Formula C9 H9 N O
Molecular Weight (g/mol) 147.17
Synonym 2-Propenamide, 3-phenyl-, (2E)-, 2-Propenamide, 3-phenyl-, (E)-, Cinnamamide, (E)- (8CI), (2E)-3-Phenyl-2-propenamide, (E)-3-Phenyl-2-propenamide, (E)-Cinnamamide, U 2422, trans-3-Phenyl-2-propenamide, trans-Cinnamamide, trans-Cinnamic acid amide, trans-Cinnamoylamine, trans-β-(Aminocarbonyl)styrene, Mitomycin Imp. A (EP)
IUPAC Name (E)-3-phenylprop-2-enamide
SMILES NC(=O)\C=C\c1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H9NO/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H2,10,11)/b7-6+
Formula Weight 147.0684
Chemical Name or Material trans-Cinnamamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.