missing translation for 'onlineSavingsMsg'
Learn More

trans-Diethyl cyclopropane-1,2-dicarboxylate, 98%, BLD Pharm™

Product Code. 30984386
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30984386 25 g
30973257 100 g
30975612 10 g
30976328 250 mg
30979964 1 g
30982402 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30984386 Supplier BLD Pharm Supplier No. BD12477925g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3999-55-1
Molecular Formula C9H14O4
MDL Number MFCD00198002
Quantity 25 g
SMILES CCOC(=O)[C@@H]1C[C@H]1C(=O)OCC
Molecular Weight (g/mol) 186.21
Percent Purity 98%
Chemical Name or Material trans-Diethyl cyclopropane-1,2-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.