missing translation for 'onlineSavingsMsg'
Learn More

trans-Methyl crotonate, 96%

Product Code. 10091950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091950 5 g Glass bottle 5g
10114320 100 g Glass bottle 100g
10124320 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091950 Supplier Thermo Scientific Acros Supplier No. 160840050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 623-43-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-43-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009287
InChI Key MCVVUJPXSBQTRZ-ONEGZZNKSA-N
Synonym methyl crotonate, e-methyl but-2-enoate, trans-methyl crotonate, methyl e-crotonate, methyl 2-butenoate, methyl trans-crotonate, 2-butenoic acid, methyl ester, 2e, trans-2-butenoic acid methyl ester, methyl trans-2-butenoate, e-crotonic acid methyl ester
PubChem CID 638132
SMILES COC(=O)\C=C\C

Specifications

Color Colorless
Density 0.9400g/mL
Boiling Point 120°C
Flash Point -1°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCO2CH3
Refractive Index 1.4230 to 1.4250
Quantity 5 g
Beilstein 02, 410
Merck Index 10, 2586
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble
IUPAC Name methyl (2E)-but-2-enoate
Formula Weight 100.12
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material trans-Methyl crotonate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • trans-Methyl crotonate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.