missing translation for 'onlineSavingsMsg'
Learn More

trans-N-Boc-4-cyano-L-proline, 97%

Product Code. 15409317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409317 250 mg 250mg
15499307 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409317 Supplier Thermo Scientific Alfa Aesar Supplier No. H52563.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 273221-94-6 | C11H16N2O4 | 240.259 g/mol

trans-N-Boc-4-cyano-L-proline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-N-Boc-4-cyano-L-proline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273221-94-6
Molecular Formula C11H16N2O4
Molecular Weight (g/mol) 240.259
MDL Number MFCD01860670
InChI Key MIVXQYMYEMUMKK-YUMQZZPRSA-N
Synonym 2s,4r-1-tert-butoxycarbonyl-4-cyanopyrrolidine-2-carboxylic acid, n-boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline, 4-cyano-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline 250mg, 4r-1-tert-butoxycarbonyl-4-cyano-l-proline
PubChem CID 17750460
IUPAC Name (2S,4R)-4-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)C#N

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 240.26
Percent Purity 97%
Chemical Name or Material trans-N-Boc-4-cyano-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.