missing translation for 'onlineSavingsMsg'
Learn More

trans-N1,N2-Dimethylcyclohexane-1,2-diamine, 97%, BLD Pharm™

Product Code. 31428790
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31428790 5 g
31426082 10 g
31427333 25 g
31414419 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31428790 Supplier BLD Pharm Supplier No. BD107615g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67579-81-1
Molecular Formula C8H18N2
MDL Number MFCD03001702
Quantity 5 g
SMILES CN[C@H]1[C@H](NC)CCCC1
Molecular Weight (g/mol) 142.24
Percent Purity 97%
Chemical Name or Material trans-N1,N2-Dimethylcyclohexane-1,2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.