Learn More
trans-Resveratrol Triacetate, TRC
Product Code. 30138510
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138510 500 mg 500mg
30133767 50 mg 50mg
30011248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138510 Supplier Toronto Research Chemicals Supplier No. TRCR150055500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42206-94-0
Molecular Formula C20H18O6
Molecular Weight (g/mol) 354.35
Synonym Triacetyl Resveratrol, 3,4',5-Triacetoxy-trans-stilbene, 3,5,4'-Tri-O-acetylresveratrol, 5-[(1E)-2-[4-(Acetyloxy)phenyl]ethenyl]-1,3-benzenediol1,3-Diacetate, Resveratrol 3,5,4'-triacetate;
IUPAC Name [4-[(E)-2-(3,5-diacetyloxyphenyl)ethenyl]phenyl] acetate
SMILES CC(=O)Oc1ccc(\C=C\c2cc(OC(=O)C)cc(OC(=O)C)c2)cc1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C20H18O6/c1-13(21)24-18-8-6-16(7-9-18)4-5-17-10-19(25-14(2)22)12-20(11-17)26-15(3)23/h4-12H,1-3H3/b5-4+
Formula Weight 354.1103
Chemical Name or Material trans-Resveratrol Triacetate