Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (T56/14), PE

Mouse Monoclonal Antibody
379.00€ - 1200.00€
Specifications
| Antigen | Transferrin Receptor |
|---|---|
| Clone | T56/14 |
| Concentration | 0.1mg/mL |
| Content And Storage | 4°C, store in dark |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10445663
![]() |
Invitrogen™
MHCD7104 |
500 μL |
379.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10343733
|
Invitrogen™
MHCD7104-4 |
2 mL |
1200.00€
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.Specifications
| Transferrin Receptor | |
| 0.1mg/mL | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| ASR | |
| PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
| P02786 | |
| 7037 | |
| Human CD71. | |
| Antibody |
| T56/14 | |
| 4°C, store in dark | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| TFRC | |
| 2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
| TFRC | |
| Primary | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.