missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ TransferTip (ES)

Product Code. 15866814
Change view
Click to view available options
Diameter (Metric) Inside:
15 μm
Unit Size:
Pack of 25
Product Code. Diameter (Metric) Inside unitSize
15866814 15 μm Pack of 25
1 options
This item is not returnable. View return policy

Product Code. 15866814

Brand: Eppendorf™ 5195000079

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eppendorf™ TransferTip (ES) is the right microcapillary for all cell manipulation applications.

  • 20° tip angle
  • Rigid flange, 1mm length
  • Heat-formed spike eases penetration
  • Sterilized via validated gamma irradiation
  • Capillary safe for optimum protection
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inside 15 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.