missing translation for 'onlineSavingsMsg'
Learn More
Learn More
transgelin/SM22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10493-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody was generated against full length transgelin 1 protein. It can cross-react with other two transgelins based on the sequence similarity.
The protein encoded by this gene is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear. Two transcript variants encoding the same protein have been found for this gene.Specifications
| transgelin/SM22 | |
| Polyclonal | |
| Unconjugated | |
| TAGLN | |
| SM22, SM22 alpha, SM22a, SM22alpha, TAGLN, TAGLN1, transgelin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21345, 25123, 6876 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P31232, P37804, Q01995 | |
| Tagln | |
| transgelin/SM22 Fusion Protein Ag0764 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction