Learn More
Fisherbrand™ Transparent Terylene Mesh PVC Tubing
Product Code. 12424080 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Diameter (Metric) Inner:
5 mm
6.5 mm
8 mm
10 mm
Unit Size:
10m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
12424080 10 mm 10m
12355869 8 mm 10m
12335869 5 mm 10m
12345869 6.5 mm 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12424080
  • ONLINE EXCLUSIVE PRICE
    25.72€
    17.07€ / 10m
    Save 8.65€
    33% Off
View all promotions
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
General purpose tubing for transfer of fluids, gases, and powdered foods
All dimensions are approximate.
  • Transparent, colorless
  • Terylene mesh reinforced wall
  • Flexible yet great inherent strength
  • Suitable for moderate pressure or vacuum applications
Specifications
Diameter (Metric) Inner 10 mm
Material PVC
Overall Length (Metric) 10 m
Thickness (Metric) 3 mm
For Use With (Application) Suitable For Moderate Pressure Or Vacuum Applications
Packaging Quantity 1 Pack
Color Transparent