Learn More
Esculape Laboratoire™ Travel First Aid Kit
Product Code. 11738542
Change view
Click to view available options
Dimensions (L x W x H):
33 x 23 x 13 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
11738542 33 x 23 x 13 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11738542 Supplier Esculape Laboratoire™ Supplier No. 7230105
  • ONLINE EXCLUSIVE PRICE
    48.30€
    26.38€ / Pack of 1
    Save 21.92€
    45% Off
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Travel first aid kit with belt loop makes this the ideal kit for quick and efficient intervention when outdoors
  • Travel kit
  • Suitable for up to 4 people
  • With belt loop
  • Weight 210 g
  • First aid guide in 6 languages
Specifications
For Use With (Application) Outdoor Use
Includes 1 survival blanket, 1 Small sterile compression bandage, 1 Large sterile compressive bandage, 2 Gauze strips 3 m x 5 cm, 1 Stretch bandage 3 mx 7 cm, 4 Sterile compresses 5 x 5 cm, 1 Roll of plaster 5 mx 2 cm, 1 Bag of 10 assorted adhesive bandages, 2 Alcoholic compresses, 2 Anti-stroke compresses with calendula, 2 Anti-mosquito compresses, 2 Sachets of anti-burn gel, 2 Sachets of bactericidal gel, 1 Pair of scissors, 1 Splinter forceps, 4 Fastening clips for bands, 1 Pair of vinyl gloves, 1 First aid guide in 6 languages
Sufficient For 4 People
Dimensions (L x W x H) 33 x 23 x 13 cm