Learn More
Skytec™ TRC715 Safety Gloves
Product Code. 30274509
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30274509 Small 10 pairs
30274598 Medium 10 pairs
30274640 Large 10 pairs
30274596 X-Large 10 pairs
30274626 2X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30274509 Supplier Skytec™ Supplier No. SKG00016ED
  • ONLINE EXCLUSIVE PRICE
    136.19€
    102.13€ / 10 pairs
    Save 34.06€
    25% Off
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Outstanding cut resistance allowing protection in high risk applications. Breathable, long lasting and flexible glove providing excellent grip in oily conditions.
  • Recycled Polyester
  • HPPE on the inside provides comfort and protection
  • Fully encapsulated steel with recycled polyester on the outside strengthens the protection against burrs and sharp edges
  • Micro foam nitrile coating gives exceptional grip while allowing warm air and moisture from inside to escape
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 4X43E, EN 388:2016 + A1:2018 4X43E
Coating Material Foam Nitrile
Color Green, Black
Cut Resistance Rating Level E
Hand Ambidextrous
Latex Free Yes
Material Steel Fiber
Product Type Cut-resistant Gloves
Size Small
EN388 Standard 4X43E
Liner Material HPPE, Recycled Polyester
Length (Metric) 230 mm