missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone Acetate, TRC

Product Code. 30454612
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454612 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454612 Supplier Toronto Research Chemicals Supplier No. TRCT719080100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone Acetate
CAS 10161-34-9
Molecular Formula C20 H24 O3
Quantity 100 mg
Synonym Estra-4,9,11-trien-3-one','17β-hydroxy-','acetate (7CI,8CI), (17β)-17-(Acetyloxy)estra-4,9,11-trien-3-one, 17β-Acetoxy-Δ4,9,11-estratrien-3-one, 17β-Acetoxyestra-4,9,11-trien-3-one, 17β-Hydroxyestra-4,9,11-trien-3-one 17-acetate, 3-Oxo-17β-acetoxyestra-4,9,11-triene, Component T-H, Component T-S, Finaplix, Finaplix H, Finaplix-S, RU 1697, Trenbolone acetate, Trienbolone acetate
InChI Formula InChI=1S/C20H24O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h9-11,17-19H,3-8H2,1-2H3/t17-,18+,19+,20+/m1/s1
SMILES CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]12C
IUPAC Name [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 312.4
Formula Weight 312.1725
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.