Learn More
Erlab Trespa™ Worktop
Product Code. 31794055 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Captair Ductless Filtering Fume Hoods
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31794055 Captair Ductless Filtering Fume Hoods Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31794055 Supplier Erlab Supplier No. I0101030001
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The worktop offers a robust and durable surface, ideal for environments requiring durability, ease of maintenance and stability in use.
  • Laboratory Worktops and Weighing Surfaces: Erlab worktops are designed to provide a durable, chemically resistant and easy-to-clean working surface for ductless enclosures, filtering storage cabinets and laboratory benches. Each surface is dimensioned to fit the corresponding Erlab equipment exactly.
  • Materials Selected for Laboratory Use
    • High-resistance materials suitable for daily contact with laboratory chemicals
    • Smooth, non-porous surface that is quick and easy to decontaminate
    • Resistant to scratching, impact and routine cleaning agents
    • Stainless Steel versions available for applications requiring maximum chemical and biological resistance
    • Segmented and weighing versions available for specific handling requirements
  • Designed for Safe Handling: The worktop geometry contributes to the containment performance of the enclosure by maintaining correct airflow at the working surface. Segmented versions allow easy removal for cleaning or for passing equipment through, while weighing surfaces provide a stable, vibration-damped supPort for precision balances.
  • Key Benefits:
    • Chemically resistant, easy-to-clean working surface.
    • Exact fit with the corresponding Erlab equipment.
    • Long service life under intensive laboratory use.
    • Options available for weighing and segmented configurations.
    • Contributes to correct airflow and containment performance.
Specifications
For Use With (Equipment) Captair Ductless Filtering Fume Hoods
Product Type Work Surface