Learn More
Emma Safety Footwear Treviso Safety Shoes
Product Code. 15998151
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998151 48 1 pair
15908151 39 1 pair
15918151 40 1 pair
15928151 41 1 pair
15938151 42 1 pair
15948151 43 1 pair
15958151 44 1 pair
15968151 45 1 pair
15978151 46 1 pair
15988151 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15998151 Supplier Emma Safety Footwear Supplier No. MM10509248
  • ONLINE EXCLUSIVE PRICE
    193.46€
    112.35€ / 1 pairs
    Save 81.11€
    41% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The foot-hugging fit provides the wearer with a naturally light and airy on-foot feel. Water resistant uppers and a woven insole make the shoes virtually penetration proof. The 100% nitrile rubber outsole repels liquids, harmful or otherwise, and provides extra grip on almost all surfaces.
  • O3-Work shoe without steel toecap with sole plate
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • Metal-free
  • Chrome-free lining
  • ESD certified
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20347:2012
Closure Type Laces
Color Black
Design Treviso
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 48