missing translation for 'onlineSavingsMsg'
Learn More

Tri(4-morpholinyl)phosphine Oxide, TRC

Product Code. 30137104
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137104 500 mg 500mg
30136527 100 mg 100mg
30003830 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137104 Supplier Toronto Research Chemicals Supplier No. TRCT896503500MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4441-12-7
Molecular Formula C12H24N3O4P
Molecular Weight (g/mol) 305.31
Synonym NSC 41250, Phosphoric Acid Trimorpholide, Phosphoric Trimorpholide, Phosphoryl Trimorpholide;
IUPAC Name 4-dimorpholin-4-ylphosphorylmorpholine
SMILES O=P(N1CCOCC1)(N2CCOCC2)N3CCOCC3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H24N3O4P/c16-20(13-1-7-17-8-2-13,14-3-9-18-10-4-14)15-5-11-19-12-6-15/h1-12H2
Formula Weight 305.1504
Chemical Name or Material Tri(4-morpholinyl)phosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.