Learn More
Tri-tert-butylphosphine, 95%, Thermo Scientific Chemicals
Product Code. 10388601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388601 1 g 1g
10522802 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10388601 Supplier Thermo Scientific Acros Supplier No. 360890010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13716-12-6
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00015006
InChI Key BWHDROKFUHTORW-UHFFFAOYSA-N
IUPAC Name tri-tert-butylphosphane
SMILES CC(C)(C)P(C(C)(C)C)C(C)(C)C
Specifications
Melting Point 30.0°C
Density 0.8200 g/mL
Boiling Point 102.0°C to 103.0°C (13.0 mmHg)
Flash Point -17°C
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula [(CH3)3C]3P
Quantity 1 g
Specific Gravity 0.82
Solubility Information Solubility aq. soln.: insoluble aq. soln..
Formula Weight 202.32
Chemical Name or Material Tri-tert-butylphosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tri-tert-butylphosphine
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Pyrophoric solid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H250-Catches fire spontaneously if exposed to air.
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.