missing translation for 'onlineSavingsMsg'
Learn More

Tri-tert-butylphosphine, 98%

Product Code. 15400987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400987 1 g 1g
15410987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400987 Supplier Thermo Scientific Alfa Aesar Supplier No. 010178.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13716-12-6 | C12H27P | 202.32 g/mol

Tri-tert-butylphosphine is used primarily in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations and other asymmetric reactions as well as transition metal coupling reactions. This reagent is also used in the preparation of other phosphines in catalytic polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13716-12-6
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00015006
InChI Key BWHDROKFUHTORW-UHFFFAOYSA-N
Synonym tri-tert-butylphosphine, tri-t-butylphosphine, tri-tert-butylphosphane, tri-tertbutylphosphine, tris 1,1-dimethylethyl phosphine, tri-tert-butylphosphin, p t-bu 3, tri-tert-butyl-phosphane, tris tert-butyl phosphine, phosphine, tris 1,1-dimethylethyl
PubChem CID 83681
IUPAC Name tri-tert-butylphosphane
SMILES CC(C)(C)P(C(C)(C)C)C(C)(C)C

Specifications

Melting Point 30°C
Color White
Boiling Point 102°C to 103°C (13 mmHg)
Flash Point -16°C (-3°F)
Odor Pungent
Linear Formula [(CH3)3C]3P
Quantity 1 g
UN Number UN2846
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water. Soluble in alcohol and organic solvents
Formula Weight 202.32
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Tri-tert-butylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.