missing translation for 'onlineSavingsMsg'
Learn More

Triacetyl Aloe-emodin (Impurity A), TRC

Product Code. 30449838
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449838 25 mg -
30449688 5 mg 5mg
30452141 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449838 Supplier Toronto Research Chemicals Supplier No. TRCT73400025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25395-11-3
Molecular Formula C21H16O8
Molecular Weight (g/mol) 396.35
Synonym 1,8-Bis(acetyloxy)-3-[(acetyloxy)methyl]-9,10-anthracenedione, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone Triacetate, Aloe-emodin Impurity A
IUPAC Name (4,5-diacetyloxy-9,10-dioxoanthracen-2-yl)methyl acetate
SMILES CC(=O)OCc1cc(OC(=O)C)c2C(=O)c3c(OC(=O)C)cccc3C(=O)c2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H16O8/c1-10(22)27-9-13-7-15-19(17(8-13)29-12(3)24)21(26)18-14(20(15)25)5-4-6-16(18)28-11(2)23/h4-8H,9H2,1-3H3
Formula Weight 396.0845
Chemical Name or Material Triacetyl Aloe-emodin (Impurity A)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.