missing translation for 'onlineSavingsMsg'
Learn More

Triazophos 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30323576
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323576 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323576 Supplier Dr. Ehrenstorfer Supplier No. DREA17650000AC1000
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triazophos
CAS 24017-47-8
Analyte or Component Names Triazophos
Shipping Condition Room Temperature
Molecular Formula C12 H16 N3 O3 P S
Concentration 1000 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym Phosphorothioic acid, O,O-diethyl O-(1-phenyl-1H-1,2,4-triazol-3-yl) ester, 1H-1,2,4-Triazol-3-ol, 1-phenyl-, O-ester with O,O-diethyl phosphorothioate (8CI), 1-Phenyl-3-(O,O-diethylthionophosphoryl)-1,2,4-triazole, HOE 2960, Hostathion, Hostathion 40EC, Hostation, Triazofos, Triazophos
InChI Formula InChI=1S/C12H16N3O3PS/c1-3-16-19(20,17-4-2)18-12-13-10-15(14-12)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3
Solution Type Single Solution
SMILES CCOP(=S)(OCC)Oc1ncn(n1)c2ccccc2
IUPAC Name diethoxy-[(1-phenyl-1,2,4-triazol-3-yl)oxy]-sulfanylidene-λ^{5}-phosphane
Solvent or Matrix Acetone
Industry Type Environmental Testing
Molecular Weight (g/mol) 313.31
Quantity 1 mL
Formula Weight 313.065 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.