missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Rabbit anti-Human, Clone: S01-9D1, Novus Biologicals™
Rabbit Monoclonal Antibody
286.00€ - 581.00€
Specifications
| Antigen | TRIB3 |
|---|---|
| Clone | S01-9D1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18355294
|
Novus Biologicals
NBP3-15103-25UL |
25 μg |
286.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18398731
|
Bio-Techne
NBP3-15103-100UL |
100 μg |
581.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIB3 Monoclonal antibody specifically detects TRIB3 in Human samples. It is validated for Western BlotSpecifications
| TRIB3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Hypoxia | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 57761 | |
| IgG | |
| Affinity purified |
| S01-9D1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C20orf97, dJ1103G7.3, neuronal cell death inducible putative kinase, Neuronal cell death-inducible putative kinase, NIPK, p65-interacting inhibitor of NF-kappaB, p65-interacting inhibitor of NF-kappa-B, SINK, SKIP3, TRB-3, TRB3chromosome 20 open reading frame 97, tribbles homolog 3, tribbles homolog 3 (Drosophila) | |
| A synthetic peptide of human TRIB3 (Uniprot#: Q96RU7) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title