missing translation for 'onlineSavingsMsg'
Learn More

Tribromoacetic acid, 97%, Thermo Scientific™

Product Code. 11339849
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339849 5 g 5g
11349849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339849 Supplier Thermo Scientific Alfa Aesar Supplier No. B24367.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-96-7
Molecular Formula C2HBr3O2
Molecular Weight (g/mol) 296.74
MDL Number MFCD00004167
InChI Key QIONYIKHPASLHO-UHFFFAOYSA-N
Synonym tribromoacetic acid, acetic acid, tribromo, tribromacetic acid, unii-71ktl99qjx, ccris 7887, 71ktl99qjx, acetic acid, 2,2,2-tribromo, snpljqbkdvidtaetp@, dsstox_cid_1668, acmc-209p0y
PubChem CID 6415
IUPAC Name 2,2,2-tribromoacetic acid
SMILES C(=O)(C(Br)(Br)Br)O

Specifications

Melting Point 130°C to 133°C
Boiling Point 244°C to 246°C
Quantity 5 g
UN Number UN3261
Beilstein 1755150
Sensitivity Light sensitive
Merck Index 14,9609
Formula Weight 296.74
Percent Purity 97%
Chemical Name or Material Tribromoacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.