missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trichloroacrylic acid, 98%

Product Code. 11443920
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443920 Supplier Alfa Aesar™ Supplier No. A12299.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2257-35-4
Melting Point 72°C to 74°C
Molecular Formula C3HCl3O2
MDL Number MFCD00015455
Quantity 1 g
UN Number UN3261
Beilstein 1754536
Synonym trichloroacrylic acid, 2,3,3-trichloroacrylic acid, trichloroprop-2-enoic acid, perchlorovinylic acid, trichlor-acrylsaure, 2-propenoic acid, 2,3,3-trichloro, acmc-1cs6k
InChI Key WMUBNWIGNSIRDH-UHFFFAOYSA-N
SMILES C(=C(Cl)Cl)(C(=O)O)Cl
IUPAC Name 2,3,3-trichloroprop-2-enoic acid
Molecular Weight (g/mol) 175.389
PubChem CID 16742
Formula Weight 175.4
Percent Purity 98%
Chemical Name or Material Trichloroacrylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.