Learn More
Trichloroethylene, 98%
Description
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane
Solubility
Soluble in water (1g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light Sensitive to light. Handle and store under inert gas. Incompatible with bases and oxidizing agents.
Specifications
Specifications
| Melting Point | -85°C |
| Density | 1.46 |
| Boiling Point | 87°C |
| Odor | Chloroform-like |
| Refractive Index | 1.476 |
| Quantity | 2500 mL |
| UN Number | UN1710 |
| Beilstein | 1736782 |
| Merck Index | 14,9639 |
| Solubility Information | Soluble in water (1g/L at 20°C). |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.