missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, ACS, 99.5% min, stab., Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 019401.AP

Quantity 500mL

 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11440500

  • 44.30€ / 500mL
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

79-01-6
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
6575
1,1,2-trichloroethene
C2HCl3
MFCD00000838
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
CHEBI:16602
C(=C(Cl)Cl)Cl

Specifications

79-01-6
87°C
9.5% min.
1.476
500mL
1736782
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
XSTXAVWGXDQKEL-UHFFFAOYSA-N
1,1,2-trichloroethene
6575
131.39
ACS
1.46
None
C2HCl3
MFCD00000838
UN1710
14,9639
Miscible with ether,alcohol,chloroformMiscible with ether,ethanol and chloroform; Very slightly miscible with water.
C(=C(Cl)Cl)Cl
131.38
CHEBI:16602
≤99.5%
Trichloroethylene, For ACS analysis
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.