missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, ACS, 99.5% min, stab., Thermo Scientific™

44.30€ - 226.00€

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440500
SDS View Documents
Thermo Scientific Alfa Aesar
019401.AP
500mL
44.30€
500mL
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11333577
SDS View Documents
Thermo Scientific Alfa Aesar
019401.K2
1L
63.80€
1 L
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11313577
SDS View Documents
Thermo Scientific Alfa Aesar
019401.M1
4 x 1L
226.00€
4 L
View alternative products
Save Up to 
»
Estimated Shipment: 08-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

79-01-6
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
6575
1,1,2-trichloroethene
C2HCl3
MFCD00000838
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
CHEBI:16602
C(=C(Cl)Cl)Cl

Specifications

79-01-6
87°C
9.5% min.
1.476
UN1710
14,9639
Miscible with ether,alcohol,chloroformMiscible with ether,ethanol and chloroform; Very slightly miscible with water.
C(=C(Cl)Cl)Cl
131.38
CHEBI:16602
≤99.5%
Trichloroethylene, For ACS analysis
1.46
None
C2HCl3
MFCD00000838
1736782
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
XSTXAVWGXDQKEL-UHFFFAOYSA-N
1,1,2-trichloroethene
6575
131.39
ACS
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H315-H319-H336

P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c

H315-H317-H319-H336-H341-H350-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE

missing translation for 'einecsNumber' : 201-167-4

missing translation for 'rtecsNumber' : KX4550000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.