missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichomonas Vaginalis 1 Antibody (1306/156), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62964AF350
This item is not returnable.
View return policy
Description
Trichomonas Vaginalis 1 Monoclonal specifically detects Trichomonas Vaginalis 1 in Protozoa samples. It is validated for ELISA.
Specifications
| Trichomonas Vaginalis 1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Protozoa | |
| IgG1 |
| ELISA | |
| 1306/156 | |
| ELISA | |
| T vaginalis, TV | |
| Native | |
| 0.1 mL | |
| Recognises T. vaginalis strain 1, an anaerobic parasitic protozoan and causative agent of the sexually transmitted disease Trichomoniasis. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction