Learn More
Triclosan Methyl-d3 Ether, TRC
Product Code. 30449350
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449350 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449350 Supplier Toronto Research Chemicals Supplier No. TRCT7742805MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TriclosanmethylD3(methoxyD3)
CAS 1020720-00-6
Molecular Formula C13 2H3 H6 Cl3 O2
Quantity 5 mg
Synonym Benzene','4-chloro-1-(2,4-dichlorophenoxy)-2-(methoxy-d3)-, Triclosan methyl D3 (methoxy D3), Triclosan Methyl-d3 Ether, Triclosan-methyl ether D3 (methoxy D3)
InChI Formula InChI=1S/C13H9Cl3O2/c1-17-13-7-9(15)3-5-12(13)18-11-4-2-8(14)6-10(11)16/h2-7H,1H3/i1D3
SMILES [2H]C([2H])([2H])Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl
IUPAC Name 2,4-dichloro-1-[4-chloro-2-(trideuteriomethoxy)phenoxy]benzene
Molecular Weight (g/mol) 306.59
Formula Weight 304.9856