missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, ∽99%, MP Biomedicals™

Product Code. 11463501 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463501 100 g 100g
11473501 250 g 250g
11493501 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11463501 Supplier MP Biomedicals Supplier No. 0210310980

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Color White
Assay Percent Range ∽ 99%
Quantity 100 g
Formula Weight 185.65
Physical Form Powder
Chemical Name or Material Triethanolamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.