missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 1,1,2-ethanetricarboxylate, 99%, Thermo Scientific™

Product Code. 11308565
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11308565

Brand: Thermo Scientific Alfa Aesar L12014.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7459-46-3
Molecular Formula C11H18O6
Molecular Weight (g/mol) 246.259
MDL Number MFCD00009154
InChI Key TVWZLLYAJDSSCJ-UHFFFAOYSA-N
Synonym triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
PubChem CID 81961
IUPAC Name triethyl ethane-1,1,2-tricarboxylate
SMILES CCOC(=O)CC(C(=O)OCC)C(=O)OCC

Specifications

Density 1.074
Boiling Point 104°C to 105°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.43
Quantity 5 g
Beilstein 1796294
Formula Weight 246.26
Percent Purity 99%
Chemical Name or Material Triethyl 1,1,2-ethanetricarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.