missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Methanetricarboxylate, TRC

Product Code. 30452535
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452535 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452535 Supplier Toronto Research Chemicals Supplier No. TRCT77668025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethyl Methanetricarboxylate
CAS 6279-86-3
Molecular Formula C10H16O6
Quantity 25 g
Synonym Diethyl (Carboethoxy)malonate, Diethyl (Ethoxycarbonyl)malonate, Diethyl 2-Ethoxycarbonylmalonate, NSC 11006, Tri(ethoxycarbonyl)methane, Tricarbethoxymethane, Tricarboethoxymethane, Triethyl Methanetricarboxylate, Tris(ethoxycarbonyl)methane
InChI Formula InChI=1S/C10H16O6/c1-4-14-8(11)7(9(12)15-5-2)10(13)16-6-3/h7H,4-6H2,1-3H3
SMILES CCOC(=O)C(C(=O)OCC)C(=O)OCC
IUPAC Name triethyl methanetricarboxylate
Molecular Weight (g/mol) 232.23
Formula Weight 232.0947
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.