missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, ≥98.0% (GC), Honeywell™

Product Code. p-7111978 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 15647040

Brand: Honeywell Research Chemicals 75530250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

≥98.0% (GC)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -76°C (literature)
Density 0.891 g/mL at 25 °C(lit.)
Boiling Point 143°C to 145°C
Flash Point 30°C
Packaging Glass Bottle
Linear Formula CH(OC2H5)3
Quantity 250 mL
UN Number UN2524
Beilstein 605384
Percent Purity >98%
Grade Performance
Chemical Name or Material Triethyl orthoformate, ≥98.0% (GC)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.