Learn More
Triethyl4-phosphonobutyrate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2327-69-7 |
| Molecular Formula | C10H21O5P |
| MDL Number | MFCD00015168 |
| Quantity | 250 mg |
| SMILES | O=C(OCC)CCCP(OCC)(OCC)=O |
| Molecular Weight (g/mol) | 252.24 |
| Percent Purity | 98% |
| Chemical Name or Material | Triethyl4-phosphonobutyrate |