missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine Hydrate, TRC

Product Code. 30452987
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452987 5 g 5g
30451758 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452987 Supplier Toronto Research Chemicals Supplier No. TRCT7766305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Triethylenetetramine Hydrate
Molecular Formula C6 H18 N4 . x(H2 O)
Quantity 5 g
Synonym N1,N2-Bis(2-aminoethyl)-1,2-ethanediamine Hydrate, 1,2-Bis(2-aminoethylamino)ethane Hydrate, 1,4,7,10-Tetraazadecane Hydrate, 1,8-Diamino-3,6-diazaoctane Hydrate, 3,6-Diazaoctane-1,8-diamine Hydrate, Ancamine TETA Hydrate, EpoxySet 145-20020 Hydrate, N,N'-Bis(2-aminoethyl)-1,2-diaminoethane Hydrate, N,N'-Bis(2-aminoethyl)-1,2-ethanediamine Hydrate, N,N'-Bis(2-aminoethyl)ethylenediamine Hydrate, NSC 443 Hydrate, RT 1AX Hydrate, Ren HY 956 Hydrate, Rutapox VE 2896 Hydrate, TECZA Hydrate, TETA Hydrate, Trien Hydrate, Trientine Hydrate, VE 2896, Z1Hydrate
InChI Formula InChI=1S/C6H18N4.H2O/c7-1-3-9-5-6-10-4-2-8;/h9-10H,1-8H2;1H2
SMILES O.NCCNCCNCCN
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine;hydrate
Molecular Weight (g/mol) 164.25
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.