missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylenetetramine (Technical Grade), TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | Trientine Hydrochloride |
| CAS | 112-24-3 |
| Molecular Formula | C6H18N4 |
| Quantity | 250 g |
| Synonym | 1,2-Ethanediamine','N1,N2-bis(2-aminoethyl)-, 1,2-Ethanediamine','N,N'-bis(2-aminoethyl)- (9CI), Triethylenetetramine (8CI), N1,N2-Bis(2-aminoethyl)-1,2-ethanediamine, 1,2-Bis(2-aminoethylamino)ethane, 1,4,7,10-Tetraazadecane, 1,8-Diamino-3,6-diazaoctane, 3,6-Diazaoctane-1,8-diamine, Ancamine TETA, Aradur HY 1300GB, Aradur HY 840-1EN, Aradur HY 951, Araldite HY 951, Araldite Hardener HY 951, DEH 24, EPH 925, Epicure 3234, Epicure 925, Epikure 3234, EpoxySet 145-20020, FD 129, H 522, HA 11, HY 951, Hardener 758, K 6, MH 91, N,N'-Bis(2-aminoethyl)-1,2-diaminoethane, N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, N,N'-Bis(2-aminoethyl)ethylenediamine, N,N'-Di(2-aminoethyl)ethylenediamine, NSC 443, RT 1AX, Ren HY 956, Ren HY 956BR, Rutapox VE 2896, TECZA, TETA, TETA (crosslinking agent), TETA-K 6, Trien, Trientine, VE 2896, Z1 |
| InChI Formula | InChI=1S/C6H18N4/c7-1-3-9-5-6-10-4-2-8/h9-10H,1-8H2 |
| SMILES | NCCNCCNCCN |
| IUPAC Name | N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
| Molecular Weight (g/mol) | 146.23 |
| Formula Weight | 146.1531 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction