missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilyl Trifluoromethanesulfonate, TRC

Product Code. 30450270
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450270 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450270 Supplier Toronto Research Chemicals Supplier No. TRCT77807010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethylsilyl Trifluoromethanesulfonate
CAS 79271-56-0
Molecular Formula C22H41F3N2O6S2Si
Quantity 10 g
Synonym 1,1,1-Trifluoromethanesulfonic acid triethylsilyl ester, Triethylsilyl triflate, Triethylsilyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid triethylsilyl ester, TESOTf
InChI Formula InChI=1S/C7H15F3O3SSi/c1-4-15(5-2,6-3)13-14(11,12)7(8,9)10/h4-6H2,1-3H3
SMILES CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
IUPAC Name triethylsilyl trifluoromethanesulfonate
Molecular Weight (g/mol) 264.34
Formula Weight 264.0463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.