Learn More
Trifluoperazine dihydrochloride, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Chemical Name or Material | Trifluoperazine hydrochloride |
| CAS | 440-17-5 |
| Analyte or Component Names | Trifluoperazine hydrochloride |
| Shipping Condition | +20°C |
| Molecular Formula | C21 H24 F3 N3 S . 2 Cl H |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Trifluoperazine hydrochloride, 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10H-phenothiazine dihydrochloride, 10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, hydrochloride (1:2)10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (9CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (6CI,8CI), 2-Trifluoromethyl-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, Eskazine, Eskazinyl, Fluoperazine, Jatroneural, Modalina, SKF 5019, Stelazine, TFP, Terfluzine, Trazine, Trifluoperazine dihydrochloride, Trifluoperazine hydrochloride, Trifluoroperazine dihydrochloride, Trifluoroperazine hydrochloride, Triftazin, Triphthasine, Triphthazine, Triphthazine dihydrochloride, Tryptazine, Tryptazine dihydrochloride, Stelazine, SKF-5019-A2 |
| InChI Formula | InChI=1S/C21H24F3N3S.2ClH/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)28-20-8-7-16(15-18(20)27)21(22,23)24;;/h2-3,5-8,15H,4,9-14H2,1H3;2*1H |
| SMILES | Cl.Cl.CN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?